CAS RN 652-39-1
IUPAC Name:
4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms:
4-fluoro-2-benzofuran-1,3-dione
Molecular Weight (g/mol):
166.11
Molecular Formula:
C8H3FO3
InChi Key:
WWJAZKZLSDRAIV-UHFFFAOYSA-N
SMILES:
FC1=CC=CC2=C1C(=O)OC2=O
1
–
3
of
3
results
1
3-Fluorophthalic anhydride, 98%
CAS: 652-39-1 Molecular Formula: C8H3FO3 Molecular Weight (g/mol): 166.11 MDL Number: MFCD00039696 InChI Key: WWJAZKZLSDRAIV-UHFFFAOYSA-N Synonym: 3-fluorophthalic anhydride,4-fluoroisobenzofuran-1,3-dione,3-fluorophthalicanhydride,4-fluoro-1,3-isobenzofurandione,1,3-isobenzofurandione, 4-fluoro,5-fluoro-isobenzofurandione,4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione,4-fluor-2-benzofuran-1,3-dion,pubchem1949,fluorophthalic anhydride PubChem CID: 69551 IUPAC Name: 4-fluoro-2-benzofuran-1,3-dione SMILES: FC1=CC=CC2=C1C(=O)OC2=O