accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 652154-10-4

IUPAC Name: bis(4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one) sulfuric acid hydrate
Synonyms: bis(4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one) sulfuric acid hydrate
Molecular Weight (g/mol): 905.03
Molecular Formula: C46H56N4O13S
InChi Key: DYCXTLLGAUPAQK-UHFFFAOYNA-N
SMILES: O.OS(O)(=O)=O.COC1=C(OC)C=C2C(=C1)N1C3C4C(CC1=O)OCC=C1CN5CCC23C5CC41.COC1=C(OC)C=C2C(=C1)N1C3C4C(CC1=O)OCC=C1CN5CCC23C5CC41
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results