accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOO

CAS RN 65295-61-6

IUPAC Name: 3-(4-fluorophenoxy)benzaldehyde
Synonyms: 3-(4-fluorophenoxy)benzaldehyde
Molecular Weight (g/mol): 216.21
Molecular Formula: C13H9FO2
InChi Key: HTBZPEDBMOXWID-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=CC(C=O)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results