accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6530-09-2

IUPAC Name: dihydrogen 1-azabicyclo[2.2.2]octan-3-amine dichloride
Synonyms: dihydrogen 1-azabicyclo[2.2.2]octan-3-amine dichloride
Molecular Weight (g/mol): 199.12
Molecular Formula: C7H16Cl2N2
InChi Key: STZHBULOYDCZET-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1CN2CCC1CC2
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results