accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NFFF

CAS RN 654-70-6

IUPAC Name: 4-amino-2-(trifluoromethyl)benzonitrile
Synonyms: 4-amino-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: PMDYLCUKSLBUHO-UHFFFAOYSA-N
SMILES: NC1=CC=C(C#N)C(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)