accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CdClCl

CAS RN 654054-66-7

IUPAC Name: dichlorocadmium hydrate
Synonyms: cadmium chloride hydrate
Molecular Weight (g/mol): 201.33
Molecular Formula: CdCl2H2O
InChi Key: OISMQLUZKQIKII-UHFFFAOYSA-L
SMILES: O.Cl[Cd]Cl
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)