accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3N(R)NH3OO

CAS RN 65427-54-5

IUPAC Name: (2R)-2,4-diazaniumylbutanoate
Synonyms: (2R)-2,4-diazaniumylbutanoate
Molecular Weight (g/mol): 119.14
Molecular Formula: C4H11N2O2
InChi Key: OGNSCSPNOLGXSM-GSVOUGTGSA-O
SMILES: [NH3+]CC[C@@H]([NH3+])C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)