accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65473-13-4

IUPAC Name: methyl[(naphthalen-1-yl)methyl]azanium
Synonyms: methyl(naphthalen-1-ylmethyl)azanium
Molecular Weight (g/mol): 172.25
Molecular Formula: C12H14N
InChi Key: MQRIUFVBEVFILS-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results