accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65473-14-5

HClH3CN(E)

CAS RN 65473-14-5

IUPAC Name: hydrogen methyl[(naphthalen-1-yl)methyl][(2E)-3-phenylprop-2-en-1-yl]amine chloride
Synonyms: hydrogen naftifine chloride
Molecular Weight (g/mol): 323.86
Molecular Formula: C21H22ClN
InChi Key: OLUNPKFOFGZHRT-YGCVIUNWSA-N
SMILES: [H+].[Cl-].CN(C\C=C\C1=CC=CC=C1)CC1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)