accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6548-09-0

IUPAC Name: (2S)-2-azaniumyl-3-(5-bromo-1H-indol-3-yl)propanoate
Synonyms: (2S)-2-ammonio-3-(5-bromo-1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 283.13
Molecular Formula: C11H11BrN2O2
InChi Key: KZDNJQUJBMDHJW-VIFPVBQESA-N
SMILES: [NH3+][C@@H](CC1=CNC2=CC=C(Br)C=C12)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)