accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOFFFNOO

CAS RN 655-07-2

IUPAC Name: 2,4-dinitro-1-(trifluoromethoxy)benzene
Synonyms: 2,4-dinitro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 252.11
Molecular Formula: C7H3F3N2O5
InChi Key: KYUUZCUXYXNPFO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(F)(F)F)C(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results