CAS RN 6554-98-9
IUPAC Name:
4-[(1E)-2-phenylethenyl]phenol
Synonyms:
trans-4-hydroxystilbene p-styrylphenol
Molecular Weight (g/mol):
196.25
Molecular Formula:
C14H12O
InChi Key:
QVLMUEOXQBUPAH-VOTSOKGWSA-N
SMILES:
OC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
trans-4-Hydroxystilbene, 98%
CAS: 6554-98-9 Molecular Formula: C14H12O Molecular Weight (g/mol): 196.25 MDL Number: MFCD00002386 InChI Key: QVLMUEOXQBUPAH-VOTSOKGWSA-N Synonym: trans-4-hydroxystilbene,4-hydroxystilbene,4-styrylphenol,4-stilbenol,e-4-stilbenol,4-2-phenylethenyl phenol,4-2-phenylvinyl phenol,4-e-2-phenylethenyl phenol,e-4-hydroxystilbene,stilben-4-ol PubChem CID: 5284650 ChEBI: CHEBI:35101 IUPAC Name: 4-[(E)-2-phenylethenyl]phenol SMILES: C1=CC=C(C=C1)C=CC2=CC=C(C=C2)O