accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOH2OH2OMn

CAS RN 6556-16-7

IUPAC Name: manganese(2+) dihydrate oxalate
Synonyms: manganese(2+) dihydrate oxalate
Molecular Weight (g/mol): 178.99
Molecular Formula: C2H4MnO6
InChi Key: HDJUVFZHZGPHCQ-UHFFFAOYSA-L
SMILES: O.O.[Mn++].[O-]C(=O)C([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results