accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3COONH(S)NH3CCH3OHO

CAS RN 65671-53-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-6-(dimethylamino)hexanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-6-(dimethylamino)hexanoic acid
Molecular Weight (g/mol): 274.36
Molecular Formula: C13H26N2O4
InChi Key: KPXRFYHPDPDJSY-UHFFFAOYNA-N
SMILES: CN(C)CCCCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results