CAS RN 65676-21-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chlorpheniramine Nitrile, TRC
CAS: 65676-21-3 Molecular Formula: C17H18ClN3 Molecular Weight (g/mol): 299.8 Synonym: α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile,α-(p-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile,Chlorpeniramine EP Impurity D; IUPAC Name: 2-(4-chlorophenyl)-4-(dimethylamino)-2-pyridin-2-ylbutanenitrile SMILES: CN(C)CCC(C#N)(C1=CC=C(C=C1)Cl)C2=CC=CC=N2