accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHNH2(S)H2NHClHCl

CAS RN 657-26-1

IUPAC Name: dihydrogen (2S)-2,6-diaminohexanoic acid dichloride
Synonyms: dihydrogen L-lysine dichloride
Molecular Weight (g/mol): 219.11
Molecular Formula: C6H16Cl2N2O2
InChi Key: JBBURJFZIMRPCZ-XRIGFGBMSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCCC[C@H](N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)