accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3P(?)

CAS RN 657408-07-6

IUPAC Name: dicyclohexyl({2',6'-dimethoxy-[1,1'-biphenyl]-2-yl})phosphane
Synonyms: sphos
Molecular Weight (g/mol): 410.54
Molecular Formula: C26H35O2P
InChi Key: VNFWTIYUKDMAOP-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results