accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClN

CAS RN 6575-09-3

IUPAC Name: 2-chloro-6-methylbenzonitrile
Synonyms: 2-chloro-6-methylbenzonitrile
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: WQWQHJNUHQEGTN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)