accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFNCl

CAS RN 65753-47-1

IUPAC Name: 2-chloro-3-(trifluoromethyl)pyridine
Synonyms: 2-chloro-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 181.54
Molecular Formula: C6H3ClF3N
InChi Key: RXATZPCCMYMPME-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CN=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)