accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSOOClCl

CAS RN 6579-54-0

IUPAC Name: 2,6-dichlorobenzene-1-sulfonyl chloride
Synonyms: 2,6-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 245.5
Molecular Formula: C6H3Cl3O2S
InChi Key: WGGKQIKICKLWGN-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1S(Cl)(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)