accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)H3NOHOO

CAS RN 658-48-0

IUPAC Name: (2S)-2-azaniumyl-3-(4-hydroxyphenyl)-2-methylpropanoate
Synonyms: AMPT
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: NHTGHBARYWONDQ-JTQLQIEISA-N
SMILES: C[C@]([NH3+])(CC1=CC=C(O)C=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results