accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65858-50-6

IUPAC Name: 5-(bromomethyl)-2,1,3-benzothiadiazole
Synonyms: 5-(bromomethyl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 229.1
Molecular Formula: C7H5BrN2S
InChi Key: JEPACAAYCVWCFI-UHFFFAOYSA-N
SMILES: BrCC1=CC2=NSN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results