accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3SCH3NH3C

CAS RN 65894-82-8

IUPAC Name: 2-(butan-2-yl)-4,5-dimethyl-2,5-dihydro-1,3-thiazole
Synonyms: 4,5-dimethyl-2-(sec-butyl)-2,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 171.3
Molecular Formula: C9H17NS
InChi Key: FLBOQJFNAYJWIA-UHFFFAOYNA-N
SMILES: CCC(C)C1SC(C)C(C)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results