accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OCH3ON(S)HO(R)N(R)CH2CH3ON(S)HO(R)N(R)CH2OHSOHOO

CAS RN 6591-63-5

IUPAC Name: bis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Synonyms: bis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular Weight (g/mol): 782.95
Molecular Formula: C40H54N4O10S
InChi Key: ZHNFLHYOFXQIOW-YKSBFIEBSA-N
SMILES: O.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results