accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSCH3OO

CAS RN 66-27-3

IUPAC Name: methyl methanesulfonate
Synonyms: methyl methanesulfonate dimethylsulfonate methylmethansulfonat MMS as-dimethyl sulfite methyl methylenesulfonate mesilate, methyl methylester kyseliny methansulfonove methylmesilate methyl mesilate
Molecular Weight (g/mol): 110.13
Molecular Formula: C2H6O3S
InChi Key: MBABOKRGFJTBAE-UHFFFAOYSA-N
SMILES: COS(C)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)