accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 660-68-4

IUPAC Name: diethylazanium chloride
Synonyms: diethylazanium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: HDITUCONWLWUJR-UHFFFAOYSA-N
SMILES: [Cl-].CC[NH2+]CC
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)