accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)H3C(S)NHO(S)HNNH2NHHNO(S)H3CCH3NHO(S)NH2OHN(S)CH3CH3OOO(S)NHO(S)HNO(S)OHNHO(S)NHNH2NHHNO(R)NHO(S)NH2NH2OSCH3CH3ONHOOO(S)NHOHNO(R)HSNHO(S)NH2OH

CAS RN 66004-57-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)