accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66018-38-0

IUPAC Name: 8,9,16-trihydroxy-14-methoxy-3-methyl-3,4,7,8,9,10-hexahydro-1H-2-benzoxacyclotetradecine-1,7-dione
Synonyms: 8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3H-2-benzoxacyclotetradecine-1,7-dione
Molecular Weight (g/mol): 362.38
Molecular Formula: C19H22O7
InChi Key: NEQZWEXWOFPKOT-UHFFFAOYNA-N
SMILES: COC1=CC(O)=C2C(=C1)C=CCC(O)C(O)C(=O)C=CCC(C)OC2=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results