accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNN

CAS RN 6602-54-6

IUPAC Name: 2-chloropyridine-3-carbonitrile
Synonyms: 2-chloropyridine-3-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: JAUPUQRPBNDMDT-UHFFFAOYSA-N
SMILES: ClC1=NC=CC=C1C#N
Molecular Weight (g/mol) 
  • (4)
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)