accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 660425-07-0

IUPAC Name: 5-chloro-2,3-dihydropyridazin-3-one
Synonyms: 5-chloro-2H-pyridazin-3-one
Molecular Weight (g/mol): 130.53
Molecular Formula: C4H3ClN2O
InChi Key: NHMLZGFOEYLZEN-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)NN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results