accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 660432-43-9

NOOFOH

CAS RN 660432-43-9

IUPAC Name: (2-fluoro-4-nitrophenyl)methanol
Synonyms: (2-fluoro-4-nitrophenyl)methanol
Molecular Weight (g/mol): 171.13
Molecular Formula: C7H6FNO3
InChi Key: XYGZVFTWIGGORD-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results