accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2OOOOCH3CH2

CAS RN 6606-59-3

IUPAC Name: 6-[(2-methylprop-2-enoyl)oxy]hexyl 2-methylprop-2-enoate
Synonyms: 6-[(2-methylprop-2-enoyl)oxy]hexyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 254.33
Molecular Formula: C14H22O4
InChi Key: SAPGBCWOQLHKKZ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCCCCCOC(=O)C(C)=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results