accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 661-69-8

IUPAC Name: hexamethyldistannane
Synonyms: hexamethyldistannane
Molecular Weight (g/mol): 327.63
Molecular Formula: C6H18Sn2
InChi Key: CCRMAATUKBYMPA-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)[Sn](C)(C)C
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)