accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 661489-23-2

IUPAC Name: 3-(4-amino-3,5-dimethylphenyl)prop-2-enenitrile hydrochloride
Synonyms: 3-(4-amino-3,5-dimethylphenyl)prop-2-enenitrile hydrochloride
Molecular Weight (g/mol): 208.69
Molecular Formula: C11H13ClN2
InChi Key: DHBOHGCHPDMVOD-UHFFFAOYSA-N
SMILES: Cl.CC1=CC(C=CC#N)=CC(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results