CAS RN 662-50-0
IUPAC Name:
heptafluorobutanamide
Synonyms:
heptafluorobutanamide
Molecular Weight (g/mol):
213.06
Molecular Formula:
C4H2F7NO
InChi Key:
FOBJABJCODOMEO-UHFFFAOYSA-N
SMILES:
NC(=O)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Heptafluorobutyramide, 97%
CAS: 662-50-0 Molecular Formula: C4H2F7NO Molecular Weight (g/mol): 213.055 MDL Number: MFCD00039773 InChI Key: FOBJABJCODOMEO-UHFFFAOYSA-N Synonym: heptafluorobutyramide,perfluorobutyramide,butanamide, 2,2,3,3,4,4,4-heptafluoro,heptafluorobutanamide,heptafluorobutyroamide,heptafluoro-n-butyramide,butyramide, 2,2,3,3,4,4,4-heptafluoro,2,2,3,3,4,4,4-heptafluorobutyramide,2,2,3,3,4,4,4-heptafluoro-butyramide,butyramide, alpha,alpha,beta,beta,gamma,gamma,gamma-heptafluoro PubChem CID: 12626 IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanamide SMILES: C(=O)(C(C(C(F)(F)F)(F)F)(F)F)N