accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66215-27-8

H2NNNH2NNHN

CAS RN 66215-27-8

IUPAC Name: N2-cyclopropyl-1,3,5-triazine-2,4,6-triamine
Synonyms: cyromazine armor larvadex neporex cyromazinum vetrazine epa pesticide chemical code 121301 azimethiphos vetrazin ciromazina
Molecular Weight (g/mol): 166.19
Molecular Formula: C6H10N6
InChi Key: LVQDKIWDGQRHTE-UHFFFAOYSA-N
SMILES: NC1=NC(NC2CC2)=NC(N)=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results