CAS RN 6622-76-0
CAS RN 6622-76-0
IUPAC Name:
methyl (2Z)-2-methylbut-2-enoate
Synonyms:
methyl tiglate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
YYJWBYNQJLBIGS-PLNGDYQASA-N
SMILES:
COC(=O)C(\C)=C/C
1
–
1
of
1
results
1
Methyl tiglate, 98%
CAS: 6622-76-0 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.14 MDL Number: MFCD00016654 InChI Key: YYJWBYNQJLBIGS-PLNGDYQASA-N Synonym: methyl tiglate,tiglic acid methyl ester,methyl 2-methylbut-2-enoate,methyl 2-methyl-2-butenoate,methyl e-2-methylcrotonate,methyl 2-methylcrotonate,methyl alpha-methylcrotonate,methyl trans-2-methylcrotonate,2-butenoic acid, 2-methyl-, methyl ester, e,crotonic acid, 2-methyl-, methyl ester, e PubChem CID: 5323652 IUPAC Name: methyl (E)-2-methylbut-2-enoate SMILES: COC(=O)C(\C)=C/C