accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHBr

CAS RN 6627-55-0

IUPAC Name: 2-bromo-4-methylphenol
Synonyms: 2-bromo-4-methylphenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: MTIDYGLTAOZOGU-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results