accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 6630-33-7

IUPAC Name: 2-bromobenzaldehyde
Synonyms: benzaldehyde, 2-bromo-
Molecular Weight (g/mol): 185.02
Molecular Formula: C7H5BrO
InChi Key: NDOPHXWIAZIXPR-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)