accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OHNOFFF

CAS RN 66332-96-5

IUPAC Name: N-[3-(propan-2-yloxy)phenyl]-2-(trifluoromethyl)benzamide
Synonyms: flutolanil
Molecular Weight (g/mol): 323.32
Molecular Formula: C17H16F3NO2
InChi Key: PTCGDEVVHUXTMP-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC(NC(=O)C2=CC=CC=C2C(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results