accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHONOO

CAS RN 6635-20-7

IUPAC Name: 4-hydroxy-3-methoxy-5-nitrobenzaldehyde
Synonyms: 5-nitrovanillin
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: ZEHYRTJBFMZHCY-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(=C1O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results