accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66356-53-4

H3CNCH3OSNH2

CAS RN 66356-53-4

IUPAC Name: 2-[({5-[(dimethylamino)methyl]furan-2-yl}methyl)sulfanyl]ethan-1-amine
Synonyms: 2-[({5-[(dimethylamino)methyl]furan-2-yl}methyl)sulfanyl]ethanamine
Molecular Weight (g/mol): 214.33
Molecular Formula: C10H18N2OS
InChi Key: JFGCGQJHMUYGLU-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=C(CSCCN)O1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results