accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOClN

CAS RN 6636-55-1

IUPAC Name: methyl 6-chloropyridine-2-carboxylate
Synonyms: methyl 6-chloropyridine-2-carboxylate
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: TWUXBVMXSBEKHA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(Cl)=N1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)