accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66361-67-9

IUPAC Name: 6-bromo-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6-bromo-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: OSDHOOBPMBLALZ-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C(=O)CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results