accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOOHOHBOHHONH2BHOOHNH2

CAS RN 66472-86-4

IUPAC Name: (3-aminophenyl)boronic acid
Synonyms: M-aminophenylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: JMZFEHDNIAQMNB-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)B(O)O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results