accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFFOHO

CAS RN 665-31-6

IUPAC Name: 2,2-difluorobutanedioic acid
Synonyms: 2,2-difluorobutanedioic acid
Molecular Weight (g/mol): 154.07
Molecular Formula: C4H4F2O4
InChi Key: ZYLFHISLYSHWRH-UHFFFAOYSA-N
SMILES: OC(=O)CC(F)(F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results