accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66534-97-2

PNPClHH

CAS RN 66534-97-2

IUPAC Name: bis[2-(diphenylphosphanyl)ethyl]amine hydrochloride
Synonyms: bis[2-(diphenylphosphanyl)ethyl]amine hydrochloride
Molecular Weight (g/mol): 477.95
Molecular Formula: C28H30ClNP2
InChi Key: AVDUVHMGGDOIQI-UHFFFAOYSA-N
SMILES: Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)