accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO(S)(S)OH(S)H3CCH3(S)HO(R)H3C(S)OHO(R)H3CO(R)H3CH2C

CAS RN 66575-29-9

IUPAC Name: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-dodecahydro-1H-naphtho[2,1-b]pyran-5-yl acetate
Synonyms: forskolin colforsinum coleonol colforsin boforsin 考福新 colforsina colforsine
Molecular Weight (g/mol): 410.51
Molecular Formula: C22H34O7
InChi Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N
SMILES: CC(=O)O[C@H]1[C@@H](O)[C@H]2C(C)(C)CC[C@H](O)[C@]2(C)[C@@]2(O)C(=O)C[C@@](C)(O[C@]12C)C=C
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (11)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (5)
  • (6)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (8)
  • (3)
  • (5)
  • (11)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (5)
  • (6)
  • (2)
  • (5)