accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CClSiCH3CH3

CAS RN 66604-31-7

IUPAC Name: (1-chlorododecyl)dimethylsilyl
Synonyms: (1-chlorododecyl)dimethylsilyl
Molecular Weight (g/mol): 261.93
Molecular Formula: C14H30ClSi
InChi Key: ZAYDZNCWLCLZDU-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(Cl)[Si](C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results