accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 666816-98-4

IUPAC Name: 8-bromo-7-(but-2-yn-1-yl)-3-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-bromo-7-(but-2-yn-1-yl)-3-methyl-1H-purine-2,6-dione
Molecular Weight (g/mol): 297.11
Molecular Formula: C10H9BrN4O2
InChi Key: HFZOBQSHTNNKFY-UHFFFAOYSA-N
SMILES: CC#CCN1C(Br)=NC2=C1C(=O)NC(=O)N2C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results